Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLEKHG2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP169120UL Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP169120UL 20 μL
NBP169100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP169120UL Supplier Novus Biologicals Supplier No. NBP16910020UL

Rabbit Polyclonal Antibody

PLEKHG2 Polyclonal specifically detects PLEKHG2 in Human samples. It is validated for Western Blot.

Specifications

Antigen PLEKHG2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Alias ARHGEF42, CLGFLJ38638, common-site lymphoma/leukemia guanine nucleotide exchange factor, DKFZp667J2325, FLJ00018, FLJ22458, PH domain-containing family G member 2, pleckstrin homology domain containing, family G (with RhoGef domain) member 2, pleckstrin homology domain-containing family G member 2
Gene Symbols PLEKHG2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to PLEKHG2 (pleckstrin homology domain containing, family G (with RhoGef domain) member 2) The peptide sequence was selected from the middle region of PLEKHG2. Peptide sequence DLTIPKHRHLLQAKNQEEKRLWIHCLQRL
Molecular Weight of Antigen 144 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64857
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.