Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLEKHM2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP181472
Description
PLEKHM2 Polyclonal specifically detects PLEKHM2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PLEKHM2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
KIAA0842RP11-169K16.1, novel RUN and PH domain-containing protein, PH domain-containing family M member 2, pleckstrin homology domain containing, family M (with RUN domain) member 2, pleckstrin homology domain-containing family M member 2, Salmonella-induced filaments A and kinesin-interacting protein, SifA (Salmonella-induced filaments A) and kinesin-interacting protein, SifA and kinesin-interacting protein, SKIP | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
PLEKHM2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:GTQEVLCQLKRDQPSPCLSSAEDSGVDEGQGSPSEMVHSSEFRVDNNHLLLLMIHVFRENEEQLFKMIRMSTGHMEGNLQLLYVLLTDCYVYLLRKGATEKPYLVEEAVSYNELDYVSV | |
0.1 mL | |
Lipid and Metabolism | |
23207 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction