Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLEKHM2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | PLEKHM2 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PLEKHM2 Polyclonal specifically detects PLEKHM2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PLEKHM2 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
KIAA0842RP11-169K16.1, novel RUN and PH domain-containing protein, PH domain-containing family M member 2, pleckstrin homology domain containing, family M (with RUN domain) member 2, pleckstrin homology domain-containing family M member 2, Salmonella-induced filaments A and kinesin-interacting protein, SifA (Salmonella-induced filaments A) and kinesin-interacting protein, SifA and kinesin-interacting protein, SKIP | |
PLEKHM2 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Lipid and Metabolism | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
23207 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:GTQEVLCQLKRDQPSPCLSSAEDSGVDEGQGSPSEMVHSSEFRVDNNHLLLLMIHVFRENEEQLFKMIRMSTGHMEGNLQLLYVLLTDCYVYLLRKGATEKPYLVEEAVSYNELDYVSV | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title