Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLET-1 Antibody (700738) [DyLight 755], Novus Biologicals™
SDP

Catalog No. NB375484 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB375484 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB375484 Supplier Novus Biologicals Supplier No. FAB6917Z

Rat Monoclonal Antibody

PLET-1 Monoclonal antibody specifically detects PLET-1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF

Specifications

Antigen PLET-1
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 700738
Conjugate DyLight 755
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias C11orf34, chromosome 11 open reading frame 34, placenta expressed transcript 1, PLET1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse PLET-1, Ala27-Leu216, Accession # Q8VEN2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 349633
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.