Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plexin A1 Antibody (708954), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB311641 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311641 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311641 Supplier Novus Biologicals Supplier No. FAB65361PECY7

Mouse Monoclonal Antibody

Plexin A1 Monoclonal antibody specifically detects Plexin A1 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Plexin A1
Applications Flow Cytometry
Classification Monoclonal
Clone 708954
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias NOV, NOVP, plexin A1, PLEXIN-A1
Host Species Mouse
Immunogen E. coli-derived recombinant human Plexin A1, Tyr964-Leu1152, Accession # Q9UIW2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Immunology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 5361
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.