Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plexin A4 Antibody (707206), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB377503 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB377503 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB377503 Supplier Novus Biologicals Supplier No. FAB58561A
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Plexin A4 Monoclonal specifically detects Plexin A4 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Plexin A4
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 707206
Conjugate APC
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias A, DKFZp434G0625, FAYV2820, FLJ35026, FLJ38287, KIAA1550DKFZp566O0546, plexin A4, plexin A4, B, plexin-A4, PLXNA4A, PLXNA4B, PRO34003
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Plexin A4, Thr24-Pro1237, Accession # Q9HCM2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91584
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.