Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plexin B1 Antibody (559830), PerCP, Novus Biologicals™
SDP

Catalog No. NB371535 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371535 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371535 Supplier Novus Biologicals Supplier No. FAB37491C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Plexin B1 Monoclonal specifically detects Plexin B1 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Plexin B1
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 559830
Conjugate PerCP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias plexin B1, SEP
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Plexin B1, Accession # O43157
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5364
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.