Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plexin B2 Antibody (3E7) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB018531 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
0.5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018531 0.025 mg
NBP14331001 0.1 mg
NBP1433105 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB018531 Supplier Novus Biologicals Supplier No. NBP1433100.025MG
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

Plexin B2 Monoclonal specifically detects Plexin B2 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Plexin B2
Applications Immunohistochemistry (Paraffin), Flow Cytometry, Immunoprecipitation, Immunohistochemistry
Classification Monoclonal
Clone 30000000
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS, pH 7.2 with 0.09% Sodium Azide
Gene Alias MM1KIAA0315dJ402G11.3, Nbla00445, PLEXB2, plexin B2, plexin-B2
Gene Symbols PLXNB2
Host Species Armenian Hamster
Immunogen The immunogen for this antibody was PlexinB2.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 23654
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.