Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plexin B2 Antibody (772417) [PerCP], Novus Biologicals™
SDP

Catalog No. NB375667 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB375667 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB375667 Supplier Novus Biologicals Supplier No. FAB6836C
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Plexin B2 Monoclonal antibody specifically detects Plexin B2 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF

Specifications

Antigen Plexin B2
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 772417
Conjugate PerCP
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation PBS
Gene Alias MM1KIAA0315dJ402G11.3, Nbla00445, PLEXB2, plexin B2, plexin-B2
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Plexin B2, Leu20-Trp1029, Accession # B2RXS4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 23654
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.