Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plexin B3 Antibody (905805), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB312189 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312189 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312189 Supplier Novus Biologicals Supplier No. FAB4958PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Plexin B3 Monoclonal antibody specifically detects Plexin B3 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Plexin B3
Applications Flow Cytometry
Classification Monoclonal
Clone 905805
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias PLEXB3, plexin B3, plexin-B3, PLEXR
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Plexin B3, His45-Gln1255, Accession # Q9ULL4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5365
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.