Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plexin D1 Antibody (752815), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB370785 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370785 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370785 Supplier Novus Biologicals Supplier No. FAB41601D
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Plexin D1 Monoclonal specifically detects Plexin D1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Plexin D1
Applications Western Blot
Classification Monoclonal
Clone 752815
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias KIAA0620MGC75353, PLEXD1, plexin D1, plexin-D1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Plexin D1, Leu47-Ala1271, Accession # Q9Y4D7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23129
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.