Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLGF Antibody (178/G10) - Azide and BSA Free
SDP

Catalog No. NB11096419 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11096419 0.1 mg
NB018062 0.02 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11096419 Supplier Novus Biologicals Supplier No. NB11096419

Mouse Monoclonal Antibody

PLGF Monoclonal specifically detects PLGF in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen PLGF
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 178/G10
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, ELISA 1-2 ug/ml, Immunoprecipitation 1-2 ug/ml
Formulation PBS with No Preservative
Gene Accession No. P49763
Gene Alias D12S1900, PGFL, placenta growth factor, placental growth factor, placental growth factor, vascular endothelial growth factor-related protein, PlGF, PlGF-2, PLGFplacental growth factor-like, SHGC-10760
Gene Symbols PIGF
Host Species Mouse
Immunogen Recombinant human PlGF-2
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 5228
Test Specificity The monoclonal will detect recombinant human PlGF-1 and PlGF-2 in WB under reducing and non-reducing conditions.
Reconstitution Centrifuge vial prior to opening. Reconstitute in sterile water to a concentration of 0.1-1.0 mg/ml. Please note the sample size of this product will be provided in reconstituted liquid form.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.