Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLGF Antibody (3B10) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018061 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018061 0.02 mg
NB11096418 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018061 Supplier Novus Biologicals Supplier No. NB110964180.02MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PLGF Monoclonal specifically detects PLGF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.

Specifications

Antigen PLGF
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen), Immunofluorescence, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 3B10
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2-5 ug/ml, Immunohistochemistry 2-10 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1-2 ug/ml, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS (pH 7.4) with No Preservative
Gene Alias D12S1900, PGFL, placenta growth factor, placental growth factor, placental growth factor, vascular endothelial growth factor-related protein, PlGF, PlGF-2, PLGFplacental growth factor-like, SHGC-10760
Gene Symbols PIGF
Host Species Mouse
Immunogen Human recombinant TNFRSF10B EC domain
Purification Method Protein G purified
Quantity 0.02 mg
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 5228
Test Specificity P1GF-2 (3B10)
Reconstitution Reconstitute the antibody with 500 ul sterile PBS and the final concentration is 200 ug/ml.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.