Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLGF Antibody (3B10) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$256.00 - $584.50
Specifications
Antigen | PLGF |
---|---|
Clone | 3B10 |
Concentration | LYOPH |
Dilution | Western Blot 2-5 ug/ml, Immunohistochemistry 2-10 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1-2 ug/ml, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen), Immunofluorescence, Immunocytochemistry, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018061
![]() |
Novus Biologicals
NB110964180.02MG |
0.02 mg |
Each for $256.00
|
|
|||||
NB11096418
![]() |
Novus Biologicals
NB11096418 |
0.1 mg |
Each for $584.50
|
|
|||||
Description
PLGF Monoclonal specifically detects PLGF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
PLGF | |
LYOPH | |
Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen), Immunofluorescence, Immunocytochemistry, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Angiogenesis | |
D12S1900, PGFL, placenta growth factor, placental growth factor, placental growth factor, vascular endothelial growth factor-related protein, PlGF, PlGF-2, PLGFplacental growth factor-like, SHGC-10760 | |
PIGF | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
3B10 | |
Western Blot 2-5 ug/ml, Immunohistochemistry 2-10 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1-2 ug/ml, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
5228 | |
IgG1 | |
Protein G purified | |
P1GF-2 (3B10) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title