Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLK1 Antibody (AZ44), Biotin, Novus Biologicals™
SDP

Catalog No. NB1002747B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002747B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002747B Supplier Novus Biologicals Supplier No. NB1002747B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PLK1 Monoclonal specifically detects PLK1 in Xenopus species samples. It is validated for ELISA, Western Blot.

Specifications

Antigen PLK1
Applications ELISA, Western Blot
Classification Monoclonal
Clone AZ44
Conjugate Biotin
Dilution Western Blot, ELISA
Formulation PBS with 0.05% Sodium Azide
Gene Alias cell cycle regulated protein kinase, EC 2.7.11, EC 2.7.11.21, PLK, PLK-1, polo (Drosophia)-like kinase, polo like kinase, polo-like kinase 1polo-like kinase (Drosophila), Serine/threonine-protein kinase 13, serine/threonine-protein kinase PLK1, STPK13
Gene Symbols PLK1
Host Species Mouse
Immunogen Baculo-expressed PLK1 from Xenopus laevis
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Mitotic Regulators, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5347
Test Specificity Plx is activated by phosphorylation. AZ44 only detects the mitotic form of Plx (Xenopus laevis) which may be due to the recognition of a phosphorylated epitope. Recommended concentration: 1 ug/ml
Target Species Xenopus species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.