Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLK1 Antibody (AZ44), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB310355 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310355 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310355 Supplier Novus Biologicals Supplier No. NB1002747MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PLK1 Monoclonal antibody specifically detects PLK1 in Xenopus samples. It is validated for Western Blot, ELISA

Specifications

Antigen PLK1
Applications Western Blot, ELISA
Classification Monoclonal
Clone AZ44
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias cell cycle regulated protein kinase, EC 2.7.11, EC 2.7.11.21, PLK, PLK-1, polo (Drosophia)-like kinase, polo like kinase, polo-like kinase 1polo-like kinase (Drosophila), Serine/threonine-protein kinase 13, serine/threonine-protein kinase PLK1, STPK13
Host Species Mouse
Immunogen Baculo-expressed PLK1 from Xenopus laevis
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Mitotic Regulators, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5347
Target Species Xenopus species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.