Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLK1 Antibody (OTI1D4), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB273472A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB273472A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB273472A48 Supplier Novus Biologicals Supplier No. NBP273472AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PLK1 Monoclonal specifically detects PLK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen PLK1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1D4
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias cell cycle regulated protein kinase, EC 2.7.11, EC 2.7.11.21, PLK, PLK-1, polo (Drosophia)-like kinase, polo like kinase, polo-like kinase 1polo-like kinase (Drosophila), Serine/threonine-protein kinase 13, serine/threonine-protein kinase PLK1, STPK13
Gene Symbols PLK1
Host Species Mouse
Immunogen Full length recombinant protein of human PLK1 (NP_005021) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Mitotic Regulators, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5347.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.