Learn More
Description
Specifications
Specifications
| Antigen | PLK1 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | OTI1D4 |
| Concentration | 2.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 2ug/500ul, Immunohistochemistry-Paraffin 1:150 |
| Gene Accession No. | P53350 |
| Gene Alias | cell cycle regulated protein kinase, EC 2.7.11, EC 2.7.11.21, PLK, PLK-1, polo (Drosophia)-like kinase, polo like kinase, polo-like kinase 1polo-like kinase (Drosophila), Serine/threonine-protein kinase 13, serine/threonine-protein kinase PLK1, STPK13 |
| Gene Symbols | PLK1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
