Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLK2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP215078 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215078 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215078 Supplier Novus Biologicals Supplier No. NBP215078
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PLK2 Polyclonal specifically detects PLK2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PLK2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.59mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS (pH 7), 20% Glycerol with 0.025% Proclin 300
Gene Accession No. Q9NYY3
Gene Alias EC 2.7.11, PLK-2, polo-like kinase 2EC 2.7.11.21, serine/threonine-protein kinase PLK2, Serine/threonine-protein kinase SNK, Serum-inducible kinase, SNKpolo-like kinase 2 (Drosophila)
Gene Symbols PLK2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PLK2. The exact sequence is proprietary.
Molecular Weight of Antigen 78 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10769
Test Specificity Human. Predicted cross-reactivity based on sequence identity: Xenopus tropicalis (91%).
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.