Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLK4 Rabbit anti-Human, DyLight 488, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB098488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB098488 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB098488 Supplier Novus Biologicals Supplier No. NBP298821G

Rabbit Polyclonal Antibody

PLK4 Polyclonal specifically detects PLK4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PLK4
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50 mM Sodium Borate
Gene Alias EC 2.7.11.21, PLK-4, polo-like kinase 4 (Drosophila), polo-like kinase 4SAKSTK18serine/threonine kinase 18, Sak, serine/threonine protein kinase SAK, Serine/threonine-protein kinase 18, serine/threonine-protein kinase PLK4, Serine/threonine-protein kinase Sak, Snk akin kinase
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human PLK4.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10733
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.