Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLOD1 Antibody, Novus Biologicals™
SDP

Catalog No. NB396679 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396679 25 μL
NBP238770 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB396679 Supplier Novus Biologicals Supplier No. NBP23877025UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

PLOD1 Polyclonal specifically detects PLOD1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.

Specifications

Antigen PLOD1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 μg/mL, Immunohistochemistry-Paraffin
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q02809
Gene Alias 2-oxoglutarate 5-dioxygenase 1, EC 1.14.11.4, Ehlers-Danlos syndrome type VI), FLJ42041, LH1, LLH, lysine hydroxylase, Lysyl hydroxylase 1, PLODLH, procollagen-lysine, procollagen-lysine 1, 2-oxoglutarate 5-dioxygenase (lysine hydroxylase, procollagen-lysine 1, 2-oxoglutarate 5-dioxygenase 1, procollagen-lysine, 2-oxoglutarate 5-dioxygenase
Gene Symbols PLOD1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: FVSLFFQRLLRLHYPQKHMRLFIHNHEQHHKAQVEEFLAQHGSEYQSVKLVGPEVRMANADARN
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5351
Test Specificity PLOD1 antibody verified on Human Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.