Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLOD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
$382.00 - $646.00
Specifications
Antigen | PLOD1 |
---|---|
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PLOD1 Polyclonal specifically detects PLOD1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
PLOD1 | |
Polyclonal | |
Rabbit | |
Human, Mouse | |
Q02809 | |
5351 | |
This antibody was developed against a recombinant protein corresponding to amino acids: FVSLFFQRLLRLHYPQKHMRLFIHNHEQHHKAQVEEFLAQHGSEYQSVKLVGPEVRMANADARN | |
Primary | |
PLOD1 antibody verified on Human Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
2-oxoglutarate 5-dioxygenase 1, EC 1.14.11.4, Ehlers-Danlos syndrome type VI), FLJ42041, LH1, LLH, lysine hydroxylase, Lysyl hydroxylase 1, PLODLH, procollagen-lysine, procollagen-lysine 1, 2-oxoglutarate 5-dioxygenase (lysine hydroxylase, procollagen-lysine 1, 2-oxoglutarate 5-dioxygenase 1, procollagen-lysine, 2-oxoglutarate 5-dioxygenase | |
PLOD1 | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title