Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLRG1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP106556 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106556 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP106556 Supplier Novus Biologicals Supplier No. NBP106556
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

PLRG1 Polyclonal specifically detects PLRG1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen PLRG1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:200 - 1:1000
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. O43660
Gene Alias MGC110980, pleiotropic regulator 1 (PRL1 homolog, Arabidopsis), PRL1Arabidopsis homolog)
Gene Symbols PLRG1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 464 and 514 of human pleiotropic regulator 1 using the numbering given in entry NP_002660.1 (GeneID 5356).
Molecular Weight of Antigen 57 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5356
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.