Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLS3 Monoclonal Antibody (C4), Invitrogen™
Mouse Monoclonal Antibody
$195.80 - $404.50
Specifications
Antigen | PLS3 |
---|---|
Clone | C4 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEMA551120
|
Invitrogen™
MA551120 |
20 μL |
Each for $195.80
|
|
|||||
FEMA551119
|
Invitrogen™
MA551119 |
200 μL |
Each for $404.50
|
|
|||||
Description
Phospholipid scramblases (PLS) translocate phospholipids bidirectionally between inner and outer leaflets of the plasma membrane and are implicated in apoptosis (1,2). Members of this family, PLS1 and PLS3 are both substrates of Protein kinase C (PKC) delta and are phosphorylated by PKC-delta during apoptosis. PKC-delta translocates to mitochondria during apoptosis and phosphorylates PLS3. Overexpression of PLS3 in the HEK293 cells enhanced apoptosis induced by UV-irradiation.Specifications
PLS3 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
AI115446; AL024105; BMND18; LOW QUALITY PROTEIN: plastin-3; MGC128570 protein; plastin 3; plastin 3 (T isoform); plastin 3 (T-isoform); plastin 3, T; plastin-3; PLS3; T fimbrin; T plastin; T-fimbrin; T-plastin | |
PLS3 | |
IgG1 κ | |
Protein A/G | |
PLS3 |
C4 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Pig, Rat | |
P13797, Q63598 | |
100158187, 5358, 81748 | |
Recombinant Human PLS3 protein, Ala379-Val630 (Accession #P13797). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title