Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLUNC Antibody, Novus Biologicals™
SDP

Catalog No. NBP226120 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226120 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP226120 Supplier Novus Biologicals Supplier No. NBP226120
Add to Cart
Add to Cart

Goat Polyclonal Antibody

PLUNC Polyclonal specifically detects PLUNC in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen PLUNC
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:64000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Alias bA49G10.5, ligand-binding protein RYA3, LPLUNC3, Lung-specific protein X, LUNXNASG, Nasopharyngeal carcinoma-related protein, Palate lung and nasal epithelium clone protein, palate, lung and nasal epithelium associated, palate, lung and nasal epithelium carcinoma associated, protein Plunc, Secretory protein in upper respiratory tracts, SPLUNC1SPURT, tracheal epithelium enriched protein, Tracheal epithelium-enriched protein, Von Ebner protein Hl
Gene Symbols BPIFA1
Host Species Goat
Immunogen Peptide with sequence C-NEVLRGLDITLVHD corresponding to C-Terminus according to NP_057667.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 51297
Test Specificity Reported variants represent identical protein: NP_057667.1, NP_001230122.1, NP_570913.1
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.