Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva PlusOne™ Reagents
Shop All Cytiva Products
Click to view available options
Quantity:
500 g
500 mL
Form:
Liquid
Solid
Description
Urea
- Provides highest resolution and the cleanest results for IEF performed under denaturing conditions
- Urea solubilizes and unfolds most proteins to their fully random conformation, with all ionizable groups exposed to solution
- 8M urea included in the 2-D sample solution
Low toxicity and environmentally safe solvent
- Used for covalently attach polyacrylamide gels to a glass surface
- Firmly attaches to the glass during staining and drying procedures
Urea:
- Molecular Weight: 60.06
- Physical state: Solid
- Color: White
- Pack of 500g
- Molecular Weight: 129.06
- Physical state: Liquid
- Color: Colorless
- Pack of 500mL
- Molecular Weight: 248.4
- Physical state: Liquid
- Color: Pale
- Pack of 25mL
IEF, PAGE, sequencing
Certifications
Suitable for DIGE
Specifications
Specifications
CAS | 2530-85-0 |
Description | PlusOne Repel-Silane ES |
Color | Colorless |
Concentration | 2% |
Product Type | Urea |
Form | Liquid |
For Use With (Application) | Used in IEF, PAGE and Sequencing |
Molecular Weight (g/mol) | 129.06 |
Quantity | 500 mL |
Storage Requirements | Dry and dark at room temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction