Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLZF Antibody (6318100), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB304173 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304173 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304173 Supplier Novus Biologicals Supplier No. FAB2944R

Mouse Monoclonal Antibody

PLZF Monoclonal antibody specifically detects PLZF in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen PLZF
Applications Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 6318100
Conjugate Alexa Fluor 647
Dilution Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias PLZFzinc finger and BTB domain-containing protein 16, zinc finger and BTB domain containing 16, Zinc finger protein 145, zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia), Zinc finger protein PLZF, ZNF145Promyelocytic leukemia zinc finger protein
Host Species Mouse
Immunogen E. coli-derived recombinant human PLZF, Met1-Gln254, Accession # Q05516
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Innate Immunity, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7704
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.