Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLZF Antibody (816421), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB363673 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363673 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363673 Supplier Novus Biologicals Supplier No. FAB29441W
Add to Cart
Add to Cart

Rat Monoclonal Antibody

PLZF Monoclonal antibody specifically detects PLZF in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining)

Specifications

Antigen PLZF
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 816421
Conjugate DyLight 650
Dilution Intracellular Staining by Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias PLZFzinc finger and BTB domain-containing protein 16, zinc finger and BTB domain containing 16, Zinc finger protein 145, zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia), Zinc finger protein PLZF, ZNF145Promyelocytic leukemia zinc finger protein
Host Species Rat
Immunogen E. coli-derived recombinant mouse PLZF, Met1-Gln254, Accession # NP_001028496
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Innate Immunity, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7704
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.