Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pma1 Antibody (40B7), Novus Biologicals™
SDP

Catalog No. NB1204645 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1204645 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1204645 Supplier Novus Biologicals Supplier No. NB1204645
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pma1 Monoclonal specifically detects Pma1 in Yeast samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Pma1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 40B7
Conjugate Unconjugated
Dilution Western Blot 1:10000, Immunohistochemistry 1:1000-1:5000, Immunocytochemistry/Immunofluorescence 1:1000-1:5000
Formulation Supplied as concentrated hybridoma cell culture media. with 5mM Sodium Azide
Gene Alias Plasma membrane ATPase 1, PMA1, Proton pump 1, YGL008C
Gene Symbols Pma1
Host Species Mouse
Immunogen Generated from a sequence of yeast Pma1 [Swiss-Prot# P05030]
Molecular Weight of Antigen 100 kDa
Purification Method Tissue culture supernatant
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 852876
Test Specificity Saccharomyces cerevisiae.
Target Species Yeast
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.