Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pma1p Monoclonal Antibody (40B7)

Catalog No. p-7246440
Change view
Click to view available options
Quantity:
250 μL
500 μL
2.5 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547323 2.5 mL
PIMA547421 500 μL
PIMA547422 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PIMA547323 Supplier Invitrogen™ Supplier No. MA547323
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pma1p Monoclonal Antibody for Western Blot, ICC/IF

Pma1p is an abundant multidomain protein of yeast which is localized in the plasma membrane. It functions as a major regulator of cytoplasmic pH by pumping protons out of the cell. It is part of the P2 subgroup of cation-transporting ATPases. Since Pma1p is a major plasma membrane protein, antibodies which bind to it are useful markers of yeast plasma membranes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pma1p
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 40B7
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Cell culture media with 5mM sodium azide
Gene PMA1
Gene Accession No. P05030
Gene Alias PMA1
Gene Symbols PMA1
Host Species Mouse
Immunogen Yeast nuclear prep.
Quantity 2.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 852876
Target Species Yeast
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.