Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMCA2 Antibody, Novus Biologicals™
SDP

Catalog No. NB1203529 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1203529 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1203529 Supplier Novus Biologicals Supplier No. NB1203529
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PMCA2 Polyclonal specifically detects PMCA2 in Human, Rat, Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen PMCA2
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry-Paraffin 1:20 - 1:200, Immunohistochemistry-Frozen 1:20 - 1:200
Formulation PBS, 1 mg/ml BSA with 0.05% Sodium Azide
Gene Accession No. Q01814, P11506
Gene Alias plasma membrane calcium-transporting ATPase 2, ATPase, Ca++ transporting, plasma membrane 2, PMCA2, PMCA2a, PMCA2i
Gene Symbols ATP2B2
Host Species Rabbit
Immunogen Synthetic Peptide: T(5) N S D F Y S K N Q R N E S S(19)
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 491
Test Specificity PMCA2 antibody detects calcium pump of the plasma membrane 2 (PMCA2 ATPase) from human and rat and porcine tissues and cells.
Target Species Human, Rat, Pig
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.