Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMCA3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159426
Description
PMCA3 Polyclonal specifically detects PMCA3 in Human samples. It is validated for Western Blot.Specifications
PMCA3 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
ATPase, Ca++ transporting, plasma membrane 3, EC 3.6.3, EC 3.6.3.8, plasma membrane calcium ATPase, Plasma membrane calcium ATPase isoform 3, Plasma membrane calcium pump isoform 3, plasma membrane calcium-transporting ATPase 3, PMCA3a, PMCA3plasma membrane calcium pump | |
Rabbit | |
Affinity purified | |
RUO | |
492 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q16720 | |
ATP2B3 | |
Synthetic peptides corresponding to ATP2B3(ATPase, Ca++ transporting, plasma membrane 3) The peptide sequence was selected from the N terminal of ATP2B3. Peptide sequence GDMANSSIEFHPKPQQQRDVPQAGGFGCTLAELRTLMELRGAEALQKIEE. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Pig: 92%; Guinea pig: 78%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction