Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMCA4 ATPase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583500
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583500 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583500 Supplier Invitrogen™ Supplier No. PA583500
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: THPEFAIEEE LPRTPLLDEE EEENPDKASK FGTRVLLLDG EVTPYANTNN NAVDCNQVQL PQSDSSLQSL ETSV Highest antigen sequence indentity to the following orthologs: Rat - 46%, Mouse - 53%.

The calcium pump of the plasma membrane, termed PMCA ATPase, pumps calcium from the cytosol to the extracellular space. This membrane-bound enzyme is related to a number of other ATPases including the SERCA ATPase and the sodium/K+ pump. There are four different genes encoding PMCA ATPase and studies have revealed 20 isoforms of the pump generated by alternate splicing of the primary gene products. mRNA distribution studies show that gene products 1 and 4 are transcribed in most tissues, however, products 2 and 3 are more tissue specific. Transcription of the splicing variants has also been found to be tissue specific. In the pancreas, where insulin secretion is calcium dependent, the beta cells only express the 4b isoform, however the alpha and gamma cells express both 4a and 4b isoforms. Studies have also shown that different splice variants have different affinities for calcium and calmodulin (Cat. # MA3-917 and MA3-918).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMCA4 ATPase
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Atp2b4
Gene Accession No. P23634
Gene Alias ATP2B2; ATP2B4; ATPase plasma membrane Ca2+ transporting 4; ATPase, Ca++ transporting, plasma membrane 4; ATPase, Ca++ transporting, plasma membrane 4 ATP synthase, H+ transporting, mitochondrial F1; ATPase, Ca++ transporting, plasma membrane 4 tv2; matrix-remodeling-associated protein 1; MXRA1; plasma membrane Ca2+-ATPases 4; plasma membrane calcium ATPase; plasma membrane calcium ATPase 4; plasma membrane calcium ATPase 4 variant b; Plasma membrane calcium ATPase isoform 4; plasma membrane calcium pump; Plasma membrane calcium pump isoform 4; plasma membrane calcium-pumping ATP-ase; plasma membrane calcium-transporting ATPase 4; PMCA4; PMCA4b; PMCA4x; sarcolemmal calcium pump
Gene Symbols Atp2b4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATP2B4. Recombinant protein control fragment (Product #RP-95778).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 493
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.