Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMEL Mouse anti-Human, Unconjugated, Clone: HMB-45, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABC1720100UG
Description
PMEL Monoclonal antibody specifically detects PMEL in Human samples. It is validated for Immunohistochemistry (Paraffin).
Specifications
PMEL | |
Monoclonal | |
Unconjugated | |
Melanocyte protein PMEL;ME20-M;ME20M;Melanocyte protein Pmel 17;Melanocytes lineage-specific antigen GP100;Melanoma-associated ME20 antigen;P1;P100;Premelanosome protein;Silver locus protein homolog | |
Mouse | |
70.25 kDa calculated. | |
100 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
HMB-45 | |
NP_008859.1 | |
PMEL, D12S53E, PMEL17 | |
Pigmented melanoma metastases extracted from human lymph nodes. | |
Protein G Purified | |
Apoptosis and Cancer | |
Clone HMB-45 is a mouse monoclonal antibody that detects human Melanocyte protein PMEL. | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction