Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMEL Mouse anti-Human, Unconjugated, Clone: HMB-45, MilliporeSigma™
Mouse Monoclonal Antibody
$189.29 - $477.80
Specifications
Antigen | PMEL |
---|---|
Clone | HMB-45 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABC17202MI
|
MilliporeSigma
MABC172025UG |
25 μg |
Each for $189.29
|
|
|||||
MABC17201MI
|
MilliporeSigma
MABC1720100UG |
100 μg |
Each for $477.80
|
|
|||||
Description
PMEL Monoclonal antibody specifically detects PMEL in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
PMEL | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
Melanocyte protein PMEL;ME20-M;ME20M;Melanocyte protein Pmel 17;Melanocytes lineage-specific antigen GP100;Melanoma-associated ME20 antigen;P1;P100;Premelanosome protein;Silver locus protein homolog | |
Pigmented melanoma metastases extracted from human lymph nodes. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
70.25 kDa calculated. |
HMB-45 | |
Monoclonal | |
Purified | |
Apoptosis and Cancer | |
NP_008859.1 | |
PMEL, D12S53E, PMEL17 | |
IgG1 κ | |
Protein G Purified | |
Clone HMB-45 is a mouse monoclonal antibody that detects human Melanocyte protein PMEL. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title