Learn More
PMEL17/SILV Antibody (PMEL/2037) - Azide and BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | PMEL17/SILV |
| Applications | Western Blot, ELISA, Immunohistochemistry (Paraffin), Peptide Array |
| Classification | Monoclonal |
| Clone | PMEL/2037 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot : 2-4 ug/mL, ELISA : 2-4 ug/mL, Immunohistochemistry-Paraffin : 1-2 ug/mL, Protein Array |
| Formulation | 10mM PBS |
| Gene Alias | D12S53EP1, gp100, ME20, ME20-M, melanocyte protein mel 17, Melanocyte protein Pmel 17, Melanocytes lineage-specific antigen GP100, Melanoma-associated ME20 antigen, melanosomal matrix protein17, PMEL17P100, premelanosome proteinME20M, SI, SIL, silver (mouse homolog) like, silver homolog (mouse), Silver locus protein homolog, silver, mouse, homolog of, SILVPmel17 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.