Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMEL17/SILV Antibody (PMEL/2038), FITC, Novus Biologicals™
SDP

Catalog No. NB380125 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB380125 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB380125 Supplier Novus Biologicals Supplier No. NBP308517F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PMEL17/SILV Monoclonal antibody specifically detects PMEL17/SILV in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen PMEL17/SILV
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone PMEL/2038
Conjugate FITC
Formulation PBS
Gene Alias D12S53EP1, gp100, ME20, ME20-M, melanocyte protein mel 17, Melanocyte protein Pmel 17, Melanocytes lineage-specific antigen GP100, Melanoma-associated ME20 antigen, melanosomal matrix protein17, PMEL17P100, premelanosome proteinME20M, SI, SIL, silver (mouse homolog) like, silver homolog (mouse), Silver locus protein homolog, silver, mouse, homolog of, SILVPmel17
Host Species Mouse
Immunogen A recombinant fragment (around aa 376-502) of human SILV protein (exact sequence is proprietary) (Uniprot: P40967)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6490
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.