Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540650
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540650 Supplier Invitrogen™ Supplier No. PA540650
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TDCYKCFYQL QPAMTQQIYD KFIAQLQTSI REEISDIKEE GNLEAVLNAL Sequence homology: Cow: 79%; Goat: 100%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 79%; Rat: 93%.

PMF1 is part of the MIS12 complex which is required for normal chromosome alignment and segregation and kinetochore formation during mitosis. It may act as a cotranscription partner of NFE2L2 involved in regulation of polyamine-induced transcription of SSAT.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMF1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PMF1
Gene Accession No. Q6P1K2
Gene Alias 2600009M07Rik; AL033286; AW060657; Est1p-like protein B (EST1B); Pmf1; PMF-1; polyamine modulated factor 1; polyamine-modulated factor 1; RP11-54H19.4; Unknown (protein for MGC:134238)
Gene Symbols PMF1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human PMF1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11243
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.