Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PML Protein Rabbit anti-Human, Mouse, Rat, Clone: 2A0N10, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP315835100UL
This item is not returnable.
View return policy
Description
PML Protein Monoclonal antibody specifically detects PML Protein in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| PML Protein | |
| Monoclonal | |
| Unconjugated | |
| MYLPP8675, promyelocytic leukemia, Promyelocytic leukemia protein, RING finger protein 71, RNF71probable transcription factor PML, TRIM19promyelocytic leukemia, inducer of, tripartite motif protein TRIM19, Tripartite motif-containing protein 19 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PML Protein (P29590). | |
| 100 μg | |
| Cancer, Zinc Finger | |
| 5371 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 2A0N10 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction