Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PML Protein Rabbit anti-Human, Mouse, Rat, Clone: 2A0N10, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | PML Protein |
---|---|
Clone | 2A0N10 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
PML Protein Monoclonal antibody specifically detects PML Protein in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
PML Protein | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Zinc Finger | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5371 | |
IgG | |
Affinity purified |
2A0N10 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
MYLPP8675, promyelocytic leukemia, Promyelocytic leukemia protein, RING finger protein 71, RNF71probable transcription factor PML, TRIM19promyelocytic leukemia, inducer of, tripartite motif protein TRIM19, Tripartite motif-containing protein 19 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PML Protein (P29590). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title