Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMM2/Phosphomannomutase 2 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB087381 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB087381 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB087381 Supplier Novus Biologicals Supplier No. NBP300351AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PMM2/Phosphomannomutase 2 Polyclonal specifically detects PMM2/Phosphomannomutase 2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PMM2/Phosphomannomutase 2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias CDG1, CDG1A, CDGS, EC 5.4.2.8, phosphomannomutase 2, PMM 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant PMM2/Phosphomannomutase 2 (O15305; Met1-Ser246).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5373
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.