Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMP22 Antibody (Hu1) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013986 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013986 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013986 Supplier Novus Biologicals Supplier No. NBP280913
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PMP22 Monoclonal specifically detects PMP22 in Human, Primate, Bovine (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PMP22
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Hu1
Concentration 0.9 mg/mL
Conjugate Unconjugated
Dilution Western Blot reported in scientific literature, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin : 1:50-1:200
Formulation Tris-Glycine, 0.15 M NaCl
Gene Alias CMT1A, CMT1E, GAS3, GAS-3DSS, growth arrest-specific 3, Growth arrest-specific protein 3, HMSNIA, HNPP, MGC20769, peripheral myelin protein 22, PMP-22, Sp110
Gene Symbols PMP22
Host Species Mouse
Immunogen Human PMP22 cDNA boosted with a 13-amino acid peptide of the 2nd extracellular domain (a.a. 120-133) conjugated to murine IgG. [Swiss-Prot# Q01453]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Glia Markers, Membrane Trafficking and Chaperones, Neuroscience, Oligodendrocyte Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5376
Test Specificity Human and Rhesus monkey. Does not react with mouse, rat, or bovine nerve protein.
Target Species Human, Primate, Bovine (Negative), Mouse (Negative), Rat (Negative)
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.