Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMP70 Rabbit anti-Human, Mouse, Rat, Clone: 3V4K3, Novus Biologicals™
SDP

Catalog No. NB167788 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167788 20 μg
NB167787 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167788 Supplier Novus Biologicals Supplier No. NBP31634820UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PMP70 Monoclonal antibody specifically detects PMP70 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen PMP70
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 3V4K3
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias 70 kDa peroxisomal membrane protein, ABC43, ATP-binding cassette, sub-family D (ALD), member 3, peroxisomal membrane protein 1 (70kD, Zellweger syndrome), Peroxisomal membrane protein-1 (70kD), PMP70ATP-binding cassette sub-family D member 3, PXMP1dJ824O18.1 (ATP-binding cassette, sub-family D (ALD), member 3 (PMP70, PXMP1)), ZWS2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 560-659 of human PMP70 (P28288).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline ABC Transporters, Cellular Markers, Lipid and Metabolism, Neuroscience, Peroxisome Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5825
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.