Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMP70 Rabbit anti-Human, Mouse, Rat, Clone: 3V4K3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
| Antigen | PMP70 |
|---|---|
| Clone | 3V4K3 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
PMP70 Monoclonal antibody specifically detects PMP70 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| PMP70 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| ABC Transporters, Cellular Markers, Lipid and Metabolism, Neuroscience, Peroxisome Markers, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 5825 | |
| IgG | |
| Affinity purified |
| 3V4K3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 70 kDa peroxisomal membrane protein, ABC43, ATP-binding cassette, sub-family D (ALD), member 3, peroxisomal membrane protein 1 (70kD, Zellweger syndrome), Peroxisomal membrane protein-1 (70kD), PMP70ATP-binding cassette sub-family D member 3, PXMP1dJ824O18.1 (ATP-binding cassette, sub-family D (ALD), member 3 (PMP70, PXMP1)), ZWS2 | |
| A synthetic peptide corresponding to a sequence within amino acids 560-659 of human PMP70 (P28288). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title