Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMS1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | PMS1 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PMS1 Polyclonal specifically detects PMS1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
PMS1 | |
Polyclonal | |
Rabbit | |
DNA Repair, Mismatch Repair, Ovarian Carcinoma Cell Markers | |
DKFZp781M0253, FLJ98259, HNPCC3, hPMS1, human homolog of yeast mutL, mismatch repair gene PMSL1, PMS1 postmeiotic segregation increased 1 (S. cerevisiae), PMS1 protein homolog 1, PMSL1DNA mismatch repair protein PMS1, postmeiotic segregation increased (S. cerevisiae) 1, rhabdomyosarcoma antigen MU-RMS-40.10B, rhabdomyosarcoma antigen MU-RMS-40.10E | |
PMS1 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
5378 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ESVLIALENLMTTCYGPLPSTNSYENNKTDVSAADIVLSKTAETDVLFNKVESSGKNYSNVDTSVIPFQNDMHNDESGKNTDDCLNHQISIGDFGYGHCSS | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title