Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMS2 Antibody (163C1251), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. N1064A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1064A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1064A94 Supplier Novus Biologicals Supplier No. NB10056554AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PMS2 Monoclonal specifically detects PMS2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PMS2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 163C1251
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DNA mismatch repair protein PMS2, EC 3.1, H_DJ0042M02.9, HNPCC4postmeiotic segregation increased (S. cerevisiae) 2, mismatch repair endonuclease PMS2, PMS1 protein homolog 2, PMS2 postmeiotic segregation increased 2 (S. cerevisiae), PMSL2PMS2CL
Gene Symbols PMS2
Host Species Mouse
Immunogen This antibody was generated by immunizing mice with a synthetic peptide corresponding to amino acids 623-639 SSLAKRIKQLHHEAQQS of human PMS2; GenBank No. NP_000526.1.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, Cell Cycle and Replication, DNA Repair, Mismatch Repair
Primary or Secondary Primary
Gene ID (Entrez) 5395.0
Test Specificity An approx. 85 kDa band should be observed.
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.