Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMS2 Antibody (163C1251) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10056554T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056554T 0.025 mg
NB10056554 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056554T Supplier Novus Biologicals Supplier No. NB10056554SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PMS2 Monoclonal specifically detects PMS2 in Human, Mouse samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen PMS2
Applications CyTOF, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 163C1251
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1ug per 10^6 cells, Immunohistochemistry 2-5ug/ml, Immunohistochemistry-Paraffin 2-5ug/ml, CyTOF-ready
Gene Accession No. P54278
Gene Alias DNA mismatch repair protein PMS2, EC 3.1, H_DJ0042M02.9, HNPCC4postmeiotic segregation increased (S. cerevisiae) 2, mismatch repair endonuclease PMS2, PMS1 protein homolog 2, PMS2 postmeiotic segregation increased 2 (S. cerevisiae), PMSL2PMS2CL
Gene Symbols PMS2
Host Species Mouse
Immunogen This antibody was generated by immunizing mice with a synthetic peptide corresponding to amino acids 623-639 SSLAKRIKQLHHEAQQS of human PMS2; GenBank No. NP_000526.1.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, DNA Repair, Mismatch Repair
Primary or Secondary Primary
Gene ID (Entrez) 5395
Test Specificity An approx. 85 kDa band should be observed.
Target Species Human, Mouse, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.