Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMS2 Antibody (163C1251) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$206.50 - $426.00

Specifications

Antigen PMS2
Clone 163C1251
Concentration 1.0 mg/mL
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1ug per 10^6 cells, Immunohistochemistry 2-5ug/ml, Immunohistochemistry-Paraffin 2-5ug/ml, CyTOF-ready
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056554T
SDP
View Documents
Novus Biologicals
NB10056554SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10056554
SDP
View Documents
Novus Biologicals
NB10056554
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

PMS2 Monoclonal specifically detects PMS2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications

Specifications

PMS2
1.0 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Unconjugated
Mouse
Cancer, Cell Cycle and Replication, DNA Repair, Mismatch Repair
P54278
5395
This antibody was generated by immunizing mice with a synthetic peptide corresponding to amino acids 623-639 SSLAKRIKQLHHEAQQS of human PMS2; GenBank No. NP_000526.1.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
163C1251
Western Blot 1-2 ug/ml, Flow Cytometry 1ug per 10^6 cells, Immunohistochemistry 2-5ug/ml, Immunohistochemistry-Paraffin 2-5ug/ml, CyTOF-ready
Monoclonal
Purified
RUO
Human, Mouse
DNA mismatch repair protein PMS2, EC 3.1, H_DJ0042M02.9, HNPCC4postmeiotic segregation increased (S. cerevisiae) 2, mismatch repair endonuclease PMS2, PMS1 protein homolog 2, PMS2 postmeiotic segregation increased 2 (S. cerevisiae), PMSL2PMS2CL
PMS2
IgG1
Protein G purified
An approx. 85 kDa band should be observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.