Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMS2 Mouse anti-Human, Mouse, Rat, Clone: 1G4E6, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | PMS2 |
---|---|
Clone | 1G4E6 |
Concentration | 1.58 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6512
![]() ![]() |
Proteintech Group Inc
660751IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
PMS1 protein homologue 2 (PMS2) is a DNA mismatch repair protein. Defects in PMS2 are a cause of hereditary non-polyposis cancer (HNPCC). Human PMS2 is an amino acid protein of 862 amino acids.Specifications
PMS2 | |
1.58 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
HNPCC4, PMS1 protein homolog 2, PMS2, PMS2CL, PMSL2 | |
Pms2 | |
IgG2a | |
Protein A | |
Antibody |
1G4E6 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P54278, P54279 | |
18861, 288479, 5395 | |
PMS2 Fusion Protein Ag12661 | |
Primary | |
-20°C | |
PMS2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title