Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMVK/phosphomevalonate kinase Antibody (003), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB333829 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333829 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333829 Supplier Novus Biologicals Supplier No. NBP290296MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PMVK/phosphomevalonate kinase Monoclonal antibody specifically detects PMVK/phosphomevalonate kinase in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen PMVK/phosphomevalonate kinase
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 003
Conjugate mFluor Violet 450 SE
Dilution ELISA, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias hPMK, HUMPMKI, phosphomevalonate kinase, PMKA, PMKase, PMKEC 2.7.4.2, PMKI
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human PMVK/phosphomevalonate kinase (Uniprot#: Q15126, Met1-Leu192)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 10654
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.